Learn More
Medchemexpress LLC Pelargonidin chloride | 134-04-3 | 98.5% | 306.70 | 1 MG
SDP
Catalog No. 5000366571
Encompass_Preferred
Pelargonidin chloride is an anthocyanidin and a scavenger of nitric oxide radical, possessing antioxidant activities. It inhibits cell viability and induces cell cycle arrest at the sub-G1 phase. Additionally, it increases the mRNA and protein expression of HO-1, NQO1, and Nrf2, and improves Aβ-induced memory and learning impairment.
  • Scavenger of nitric oxide radical
  • Antioxidant activities
  • Inhibits cell viability
  • Induces cell cycle arrest at sub-G1 phase
  • Increases mRNA and protein expression of HO-1, NQO1, Nrf2
  • Improves Aβ-induced memory and learning impairment
Catalog No. 50-003-66571 Supplier Medchemexpress LLC Supplier No. HYW0113701MG
Add to Cart
Add to Cart
Pelargonidin chloride is an anthocyanidin and a scavenger of nitric oxide radical, possessing antioxidant activities. It inhibits cell viability and induces cell cycle arrest at the sub-G1 phase. Additionally, it increases the mRNA and protein expression of HO-1, NQO1, and Nrf2, and improves Aβ-induced memory and learning impairment.
  • Scavenger of nitric oxide radical
  • Antioxidant activities
  • Inhibits cell viability
  • Induces cell cycle arrest at sub-G1 phase
  • Increases mRNA and protein expression of HO-1, NQO1, Nrf2
  • Improves Aβ-induced memory and learning impairment