Learn More
Medchemexpress LLC Pelargonidin (chloride) | 134-04-3 | 99.1% | 306.70 | 1 ML
SDP
Catalog No. 5000366570
Encompass_Preferred
Pelargonidin chloride is an anthocyanidin that acts as a scavenger of nitric oxide radicals and possesses antioxidant activities. It inhibits cell viability and induces cell cycle arrest at the sub-G1 phase. Additionally, Pelargonidin chloride increases the mRNA and protein expression of HO-1, NQO1, and Nrf2, and has been shown to improve Aβ-induced memory and learning impairment. It is intended for research use only and is not sold to patients.
  • Antioxidant activities: acts as a scavenger of nitric oxide radicals.
  • Cell viability inhibition: inhibits cell viability in a time- and dose-dependent manner in JB6 P+ cells.
  • Cell cycle arrest: induces cell cycle arrest at the sub-G1 phase in a dose-dependent manner.
  • Gene expression modulation: increases the mRNA and protein expression of HO-1, NQO1, and Nrf2.
  • Cognitive improvement: improves Aβ-induced memory and learning impairment.
Catalog No. 50-003-66570 Supplier Medchemexpress LLC Supplier No. HYW01137010MM/1ML
Add to Cart
Add to Cart
Pelargonidin chloride is an anthocyanidin that acts as a scavenger of nitric oxide radicals and possesses antioxidant activities. It inhibits cell viability and induces cell cycle arrest at the sub-G1 phase. Additionally, Pelargonidin chloride increases the mRNA and protein expression of HO-1, NQO1, and Nrf2, and has been shown to improve Aβ-induced memory and learning impairment. It is intended for research use only and is not sold to patients.
  • Antioxidant activities: acts as a scavenger of nitric oxide radicals.
  • Cell viability inhibition: inhibits cell viability in a time- and dose-dependent manner in JB6 P+ cells.
  • Cell cycle arrest: induces cell cycle arrest at the sub-G1 phase in a dose-dependent manner.
  • Gene expression modulation: increases the mRNA and protein expression of HO-1, NQO1, and Nrf2.
  • Cognitive improvement: improves Aβ-induced memory and learning impairment.