Learn More
Medchemexpress LLC Pelargonidin (chloride) | 134-04-3 | 99.1% | 306.70 | 5 MG
SDP
Catalog No. 5000455065
Encompass_Preferred
Pelargonidin chloride is an anthocyanidin that functions as a scavenger of nitric oxide radicals and possesses antioxidant activities. It has been shown to inhibit cell viability and induce cell cycle arrest at the sub-G1 phase. Additionally, it enhances the mRNA and protein expression of HO-1, NQO1, and Nrf2, and improves Aβ-induced memory and learning impairment. It is for research use only.
  • Scavenges nitric oxide radicals
  • Possesses antioxidant activities
  • Inhibits cell viability
  • Induces cell cycle arrest at sub-G1 phase
  • Increases mRNA and protein expression of HO-1, NQO1, and Nrf2
  • Improves Aβ-induced memory and learning impairment
Catalog No. 50-004-55065 Supplier Medchemexpress LLC Supplier No. HYW0113705MG
Pelargonidin chloride is an anthocyanidin that functions as a scavenger of nitric oxide radicals and possesses antioxidant activities. It has been shown to inhibit cell viability and induce cell cycle arrest at the sub-G1 phase. Additionally, it enhances the mRNA and protein expression of HO-1, NQO1, and Nrf2, and improves Aβ-induced memory and learning impairment. It is for research use only.
  • Scavenges nitric oxide radicals
  • Possesses antioxidant activities
  • Inhibits cell viability
  • Induces cell cycle arrest at sub-G1 phase
  • Increases mRNA and protein expression of HO-1, NQO1, and Nrf2
  • Improves Aβ-induced memory and learning impairment