Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Pelargonidin chloride

Catalog No. P16595MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16595MG 5 mg
P165910MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16595MG Supplier Sigma Aldrich Supplier No. P16595MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Involved in pharmacological studies investigating the use of flavonoids as inhibitors of cd38antioxidant and anthocyanidin:
  • Found in various natural sources evaluated for inhibitory effects on colon and liver cancer cells
  • Used as hyroperoxide and hydrogen peroxide scavenging substance
  • Used to study relationship between structure, antioxidant capacity and redox potentials
  • Studied to determine the mechanism of radical-scavengingantioxidant and anthocyanidin.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. p1659.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin

Specifications

CAS 134-04-3
Molecular Formula C15H11ClO5
Linear Formula C15H11ClO5
MDL Number MFCD00017589
Quantity 5 mg
Synonym 3,4 ,5,7-Tetrahydroxyflavylium chloride; 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-1-benzopyrylium chloride; Pelargonidol chloride
Molecular Weight (g/mol) 306.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.