Learn More
Medchemexpress LLC Pelcitoclax | 1619923-36-2 | 99.7% | 1281.85 | 10 MG
SDP
Catalog No. 5000344125
Encompass_Preferred
Pelcitoclax (APG-1252) is a potent Bcl-2/Bcl-xl inhibitor with antineoplastic and pro-apoptotic effects. It is for research use only. In vitro, APG-1252 changes to a reactive metabolite named APG-1252-M1, which has remarkable antitumor effects in acute myeloid leukemia. In vivo, Pelcitoclax (APG-1252) treatment inhibits xenograft tumor growth.
  • Potent Bcl-2/Bcl-xl inhibitor
  • Antineoplastic and pro-apoptotic effects
  • Changes to reactive metabolite APG-1252-M1 with antitumor effects in acute myeloid leukemia
  • Inhibits xenograft tumor growth in vivo
  • Solid appearance
  • White to off-white color
Catalog No. 50-003-44125 Supplier Medchemexpress LLC Supplier No. HY10918510MG
Pelcitoclax (APG-1252) is a potent Bcl-2/Bcl-xl inhibitor with antineoplastic and pro-apoptotic effects. It is for research use only. In vitro, APG-1252 changes to a reactive metabolite named APG-1252-M1, which has remarkable antitumor effects in acute myeloid leukemia. In vivo, Pelcitoclax (APG-1252) treatment inhibits xenograft tumor growth.
  • Potent Bcl-2/Bcl-xl inhibitor
  • Antineoplastic and pro-apoptotic effects
  • Changes to reactive metabolite APG-1252-M1 with antitumor effects in acute myeloid leukemia
  • Inhibits xenograft tumor growth in vivo
  • Solid appearance
  • White to off-white color