Learn More
Medchemexpress LLC HY-109185 1mg , Pelcitoclax CAS:1619923-36-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1091851MG
Encompass_Preferred
HY-109185 1mg Pelcitoclax CAS: 1619923-36-2 Pelcitoclax (APG-1252) is a potent Bcl-2/Bcl-xl inhibitor with antineoplastic and pro-apoptotic effects. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9224