Learn More
Medchemexpress LLC Pelcitoclax (APG-1252) | 1619923-36-2 | 99.7% | 1281.85 g·mol⁻¹ | C57H66ClF4N6O11PS4 | 5 MG
SDP
Catalog No. 5000264009
Encompass_Preferred
Pelcitoclax is a small-molecule dual inhibitor of Bcl-2 and Bcl-xL used in oncology research. It induces apoptosis and has demonstrated antitumor activity in preclinical models, and is supplied with characterized molecular properties and high analytical purity for research applications.
  • Dual Bcl-2 and Bcl-xL inhibition mechanism
  • Promotes apoptosis in tumor cells
  • Demonstrated antitumor activity in preclinical studies
  • High analytical purity for consistent results
  • Available as solid and ready-to-use DMSO solution formats
Catalog No. 50-002-64009 Supplier Medchemexpress LLC Supplier No. HY1091855MG
Pelcitoclax is a small-molecule dual inhibitor of Bcl-2 and Bcl-xL used in oncology research. It induces apoptosis and has demonstrated antitumor activity in preclinical models, and is supplied with characterized molecular properties and high analytical purity for research applications.
  • Dual Bcl-2 and Bcl-xL inhibition mechanism
  • Promotes apoptosis in tumor cells
  • Demonstrated antitumor activity in preclinical studies
  • High analytical purity for consistent results
  • Available as solid and ready-to-use DMSO solution formats