Learn More
Ted Pella Inc PELCO Center-Marked Grids, 200 mesh, 3.0mm O.D., Copper, 100/vial
SDP
Catalog No. NC0643980
Encompass
The PELCO Grids are usually somewhat thicker than similar TEM grids made by electro plating processes which makes them stiffer and easier to handle without the danger of bending, but still maintain a good definition of the grid bars. The wide rims (350-400µm) on the PELCO Grids provide additional stiffness and minimize the potential of interfering with the sample when picking up and handling the grids with tweezers
Catalog No. NC0643980 Supplier Ted Pella Inc Supplier No. 1GC200
The PELCO Grids are usually somewhat thicker than similar TEM grids made by electro plating processes which makes them stiffer and easier to handle without the danger of bending, but still maintain a good definition of the grid bars. The wide rims (350-400µm) on the PELCO Grids provide additional stiffness and minimize the potential of interfering with the sample when picking up and handling the grids with tweezers