Learn More
Ted Pella Inc PELCO Clear Polypropylene Storage Box with Dividers
SDP
Catalog No. NC1189793
Encompass
The PELCO Clear Polypropylene Storage Box is ideal for storing both samples and metallographic mounts. This box will accommodate metallographic mounts up to 2" or 50mm diameter. Comes complete with 16 dividers which can be set at 1/16" (1.6mm) intervals to create exact fitting compartments for your sample and/or mounts. The sturdy box has hinges and latches and is made from clear impact polyprolylene.
Catalog No. NC1189793 Supplier Ted Pella Inc Supplier No. 139900
The PELCO Clear Polypropylene Storage Box is ideal for storing both samples and metallographic mounts. This box will accommodate metallographic mounts up to 2" or 50mm diameter. Comes complete with 16 dividers which can be set at 1/16" (1.6mm) intervals to create exact fitting compartments for your sample and/or mounts. The sturdy box has hinges and latches and is made from clear impact polyprolylene.