Learn More
Ted Pella Inc PELCO COLLOIDAL SILVER 30G
SDP
Catalog No. NC9258915
Encompass
PELCO Colloidal Silver, Paint, 200 deg.C, Silver 60%, lacquer binder, Medium mechanical strength, 30g
Catalog No. NC9258915 Supplier Ted Pella Inc Supplier No. 16031
PELCO Colloidal Silver, Paint, 200 deg.C, Silver 60%, lacquer binder, Medium mechanical strength, 30g