Learn More
Ted Pella Inc PELCO Conductive Graphite, Isopropanol base, 30g
SDP
Supplier:  Ted Pella Inc 16053
Encompass
Conductive graphite paint, primarily designed for SEM sample preparation. The graphite flakes, with an average size of 1µm, are bonded by a cellulose resin with isopropanol as diluent. Good bonding properties on most materials. Graphite content is 20%. Ideal to mount samples on sample stubs or to make a conductive path to the sample surface. Let fully dry before using in the SEM. Maximum service temperature: 200° C.
Catalog No. NC9296112