Learn More
Ted Pella Inc PELCO Cryo-Embedding Compound, 4 oz.
SDP
Catalog No. NC0611550
Encompass
PELCO Cryo-Embedding Compound is a water soluble glycol medium for use in embedding for frozen sections. This low viscosity compound assures reliable freezing and smooth sections.
This item is not returnable. View return policy
Catalog No. NC0611550 Supplier Ted Pella Inc Supplier No. 27300
PELCO Cryo-Embedding Compound is a water soluble glycol medium for use in embedding for frozen sections. This low viscosity compound assures reliable freezing and smooth sections.