Learn More
Ted Pella Inc PELCO Flat Embedding Mold, Translucent
SDP
Catalog No. NC0561014
Encompass
All PELCO Silicone Rubber Molds are made from the highest quality silicone rubber available. With care they will last a long time. After polymerization in a 60° C oven, let molds cool before removing blocks. This precaution will increase the lifespan of the mold. Using silicone rubber with Spurr's Resin will significantly reduce the lifespan of the molds. Maximum temperature: 218°C (425°F).
Catalog No. NC0561014 Supplier Ted Pella Inc Supplier No. 10504
All PELCO Silicone Rubber Molds are made from the highest quality silicone rubber available. With care they will last a long time. After polymerization in a 60° C oven, let molds cool before removing blocks. This precaution will increase the lifespan of the mold. Using silicone rubber with Spurr's Resin will significantly reduce the lifespan of the molds. Maximum temperature: 218°C (425°F).