Learn More
Ted Pella Inc PELCO FLAT EMBEDDING MOLDS
SDP
Catalog No. NC0272843
Encompass
Alphanumeric self-impression cavities, pretrimmed end. available in blue, white, or translucent silicone rubber. Maximum temperature: 218a Deg. C (425a Deg. F). all PELCOa Silicone Rubber Molds are made from the highest quality silicone rubber available. With care they will last a long time. after polymerization in a 60a Deg. C oven, let molds cool before removing blocks. This precaution will increase the lifespan of the mold. Using silicone rubber with Spurr's Resin will significantly reduce the lifespan of the molds.
Catalog No. NC0272843 Supplier Ted Pella Inc Supplier No. 105
Alphanumeric self-impression cavities, pretrimmed end. available in blue, white, or translucent silicone rubber. Maximum temperature: 218a Deg. C (425a Deg. F). all PELCOa Silicone Rubber Molds are made from the highest quality silicone rubber available. With care they will last a long time. after polymerization in a 60a Deg. C oven, let molds cool before removing blocks. This precaution will increase the lifespan of the mold. Using silicone rubber with Spurr's Resin will significantly reduce the lifespan of the molds.