Learn More
Ted Pella Inc PELCO TEM Grid Storage Box
SDP
Catalog No. NC1734089
Encompass
Widely used compact grid box for 50 standard 3.05mm TEM grids with smoothly sliding clear plastic cover is matched to a tough, clean, self-lubricating base. The diamond-shaped grid compartments are designed to minimize awkward wrist or hand movements. The depth of the holes restricts grid movement when transported. The materials selected for the PELCO Grid Box are static dissipative to avoid static charges on the grids. Dimensions: 77.5 x 41 x 9mm.
Catalog No. NC1734089 Supplier Ted Pella Inc Supplier No. 160
Widely used compact grid box for 50 standard 3.05mm TEM grids with smoothly sliding clear plastic cover is matched to a tough, clean, self-lubricating base. The diamond-shaped grid compartments are designed to minimize awkward wrist or hand movements. The depth of the holes restricts grid movement when transported. The materials selected for the PELCO Grid Box are static dissipative to avoid static charges on the grids. Dimensions: 77.5 x 41 x 9mm.