Learn More
Ted Pella Inc PELCO Image Tabs™, Carbon Conductive Tabs, 12mm OD, 100/pk
SDP
Catalog No. NC9967923
Encompass
The thicker PELCO Image Tabs™ is a stiff 260µm (10mil) thick smooth conductive tab. Though not as conductive or sticky as the current 5mil PELCO Tabs™, PELCO Image Tabs™ are suitable as a photographic background and are repositionable. This tab is also suitable for Jet Scan applications where the tab must be removed and archived. The PELCO Image Tab's™ surface is less prone to bubbling and cracking when metal coated under vacuum and thus better than the 125µm (5mil) PELCO Tab™ as a photographic background for small particles like pollen and insect parts. Both sides of the 200µm thick conductive polycarbonate base have 30µm thick conductive adhesive. Total thickness is 260µm with a liner on both sides. Protection for handling, storage and shipping is provided on one side by the transparent liner sheet and on the other side by the white liner cover.
Catalog No. NC9967923 Supplier Ted Pella Inc Supplier No. 1608420
The thicker PELCO Image Tabs™ is a stiff 260µm (10mil) thick smooth conductive tab. Though not as conductive or sticky as the current 5mil PELCO Tabs™, PELCO Image Tabs™ are suitable as a photographic background and are repositionable. This tab is also suitable for Jet Scan applications where the tab must be removed and archived. The PELCO Image Tab's™ surface is less prone to bubbling and cracking when metal coated under vacuum and thus better than the 125µm (5mil) PELCO Tab™ as a photographic background for small particles like pollen and insect parts. Both sides of the 200µm thick conductive polycarbonate base have 30µm thick conductive adhesive. Total thickness is 260µm with a liner on both sides. Protection for handling, storage and shipping is provided on one side by the transparent liner sheet and on the other side by the white liner cover.