Learn More
Ted Pella Inc PELCO Pro C1000 Cyanoacrylate Glue, 20ml
SDP
Catalog No. NC0427986
Encompass
PELCO Pro C1000 CA glue has a high viscosity and a longer curing time. Ideally suited when more careful positioning is needed, surfaces are not smooth or for gaps up to 0.008” (200µm). Curing time 10-15 seconds.
Catalog No. NC0427986 Supplier Ted Pella Inc Supplier No. 14477
PELCO Pro C1000 CA glue has a high viscosity and a longer curing time. Ideally suited when more careful positioning is needed, surfaces are not smooth or for gaps up to 0.008” (200µm). Curing time 10-15 seconds.