Learn More
Ted Pella Inc PELCO Pro High Precision, extra fine tips, superior finish
SDP
Catalog No. NC0440008
Encompass
The PELCO Pro High Precision tweezers are made from a non-magnetic, low carbon stainless steel. The tweezers exhibit good corrosion resistance to most chemicals, salts and acids. Typical applications for these tweezers include microscopy preparation, sample handling, precision manufacturing, electronic industry, watch making and general laboratory and medical applications. Although the tweezers are made from a non-magnetic alloy, they are not recommended for use where strong magnetic fields are present.
Catalog No. NC0440008 Supplier Ted Pella Inc Supplier No. 53675NM
The PELCO Pro High Precision tweezers are made from a non-magnetic, low carbon stainless steel. The tweezers exhibit good corrosion resistance to most chemicals, salts and acids. Typical applications for these tweezers include microscopy preparation, sample handling, precision manufacturing, electronic industry, watch making and general laboratory and medical applications. Although the tweezers are made from a non-magnetic alloy, they are not recommended for use where strong magnetic fields are present.