Learn More
Ted Pella Inc PELCO Pro Wafer Tweezers, 4 Prong Tip
SDP
Catalog No. NC1090705
Encompass
The PELCO Pro Wafer Tweezers are specially designed with flat tips for safely gripping and handling Silicon Wafers. Manufactured froom non-magnetic stainless steel.
Catalog No. NC1090705 Supplier Ted Pella Inc Supplier No. 5795
The PELCO Pro Wafer Tweezers are specially designed with flat tips for safely gripping and handling Silicon Wafers. Manufactured froom non-magnetic stainless steel.