Learn More
Ted Pella Inc PELCO Tabs™, Carbon Conductive Tabs, 6mm OD
SDP
Catalog No. NC0107425
Encompass
The simplicity of application and smooth, clean surfaces of PELCO Tabs™ Carbon Conductive Tabs are a significant improvement compared to many of the other common adhesives that have been used in SEM mounting. Both tab sides have a thick conductive adhesive (conductive inner tab is 35µm and the adhesive is 45µm on each side for a total thickness of 125µm [5 mils]) with a liner on both sides, a transparent liner and white liner respectively. They are ready for immediate use. The top protective liner does not have to be removed until the sample is ready to be mounted. This reduces possible contamination. Outgassing is negligible.
Catalog No. NC0107425 Supplier Ted Pella Inc Supplier No. 160846
The simplicity of application and smooth, clean surfaces of PELCO Tabs™ Carbon Conductive Tabs are a significant improvement compared to many of the other common adhesives that have been used in SEM mounting. Both tab sides have a thick conductive adhesive (conductive inner tab is 35µm and the adhesive is 45µm on each side for a total thickness of 125µm [5 mils]) with a liner on both sides, a transparent liner and white liner respectively. They are ready for immediate use. The top protective liner does not have to be removed until the sample is ready to be mounted. This reduces possible contamination. Outgassing is negligible.