Learn More
Ted Pella Inc PELCO Ultra 19 Vacuum Pump Oil, 1 L
SDP
Catalog No. NC1416084
Encompass
Premium quality vacuum pump oil for single and dual stage rotary pumps. Extra refined for increased lubrication, enhanced sealing and reduced back streaming. Excellent oil for rotary backing pumps on high vacuum systems, electron microscopes and mass spectrometers.
Catalog No. NC1416084 Supplier Ted Pella Inc Supplier No. 89131
Premium quality vacuum pump oil for single and dual stage rotary pumps. Extra refined for increased lubrication, enhanced sealing and reduced back streaming. Excellent oil for rotary backing pumps on high vacuum systems, electron microscopes and mass spectrometers.