Learn More
TARGETMOL CHEMICALS INC PELDESINE 5MG
SDP
Catalog No. 502581456
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Peldesine (BCX 34) is an effective competitive reversible and orally active inhibitor of purine nucleoside phosphorylase. Peldesine inhibits T-cell proliferation with an IC50 of 800 nM. Peldesine can be used in research on cutaneous T-cell lymphoma psoriasis and HIV infection. purity: 99%
Catalog No. 50-258-1456 Supplier TARGETMOL CHEMICALS INC Supplier No. T123995MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Peldesine (BCX 34) is an effective competitive reversible and orally active inhibitor of purine nucleoside phosphorylase. Peldesine inhibits T-cell proliferation with an IC50 of 800 nM. Peldesine can be used in research on cutaneous T-cell lymphoma psoriasis and HIV infection. purity: 99%