Learn More
Medchemexpress LLC Peldesine dihydrochloride | 2772702-10-8 | MFCD32878297 | 99.3% | 314.17 g/mol | C12H13Cl2N5O | 10 MG
SDP
Catalog No. 5000239857
Encompass_Preferred
Peldesine dihydrochloride is the dihydrochloride salt of a potent purine nucleoside phosphorylase (PNP) inhibitor used in preclinical biochemical and pharmacological research. It exhibits low-nanomolar PNP inhibition across species and suppresses T-cell proliferation, supporting studies in T-cell lymphoma, psoriasis, and HIV-related immunology.
  • Potent PNP inhibition with low-nanomolar IC50s for human, rat, and mouse.
  • Inhibits T-cell proliferation (IC50 ~800 nM), useful for immunology studies.
  • High purity (~99.3%) suitable for research applications.
  • Molecular weight 314.17 g/mol and solid, white to off-white appearance.
  • Available in small research pack sizes and as DMSO solution formats for assays.
Catalog No. 50-002-39857 Supplier Medchemexpress LLC Supplier No. HY106934A10MG
Peldesine dihydrochloride is the dihydrochloride salt of a potent purine nucleoside phosphorylase (PNP) inhibitor used in preclinical biochemical and pharmacological research. It exhibits low-nanomolar PNP inhibition across species and suppresses T-cell proliferation, supporting studies in T-cell lymphoma, psoriasis, and HIV-related immunology.
  • Potent PNP inhibition with low-nanomolar IC50s for human, rat, and mouse.
  • Inhibits T-cell proliferation (IC50 ~800 nM), useful for immunology studies.
  • High purity (~99.3%) suitable for research applications.
  • Molecular weight 314.17 g/mol and solid, white to off-white appearance.
  • Available in small research pack sizes and as DMSO solution formats for assays.