Learn More
MSC Pelican Clamshell Hard Case, 28.20 x 19.66 x 17.63 in, Watertight
Catalog No. 5000534481
Encompass
This clamshell hard case is engineered from polypropylene, offering superior protection for sensitive equipment. It features a watertight, dustproof, and crush-proof design, making it suitable for demanding environments. With internal dimensions of 28.20 in length, 19.66 in width, and 17.63 in depth, it provides ample space while ensuring contents are secure and protected.
  • Watertight and dustproof construction.
  • Crush proof, solid wall design.
  • Padlockable for secure storage.
  • Features folding handle and polyurethane wheels for easy transport.
  • Automatic pressure equalization valve.
  • O-ring seal for reliable protection.
  • Withstands extreme temperatures from -40°C to 99°C.
  • Meets STANAG 4280, IP67, and DEF STAN 81-41 standards.
Catalog No. 50-005-34481 Supplier MSC Supplier No. 70461314
This clamshell hard case is engineered from polypropylene, offering superior protection for sensitive equipment. It features a watertight, dustproof, and crush-proof design, making it suitable for demanding environments. With internal dimensions of 28.20 in length, 19.66 in width, and 17.63 in depth, it provides ample space while ensuring contents are secure and protected.
  • Watertight and dustproof construction.
  • Crush proof, solid wall design.
  • Padlockable for secure storage.
  • Features folding handle and polyurethane wheels for easy transport.
  • Automatic pressure equalization valve.
  • O-ring seal for reliable protection.
  • Withstands extreme temperatures from -40°C to 99°C.
  • Meets STANAG 4280, IP67, and DEF STAN 81-41 standards.