Learn More
MSC Pelican Clamshell Hard Case, Layered Foam, 20-15/32 in Wide, Black
Catalog No. 5000515703
Encompass
This clamshell hard case provides robust protection for equipment. Constructed from durable polypropylene with layered foam, it measures 20-15/32 inches wide and is black. It's crush-proof, watertight, dust-proof, and IP67 rated. Features include an automatic pressure equalization valve and polyurethane wheels for easy transport.
  • Layered foam for customizable protection.
  • Padlockable design for secure storage.
  • Folding handles enable easy carrying.
  • Durable polypropylene construction.
  • IP67 rated for water and dust resistance.
  • Withstands -40°F to 210°F.
  • Crush-proof, solid wall design.
  • O-ring seal ensures watertight integrity.
  • Automatic pressure equalization valve.
  • Lightweight with polyurethane wheels.
Catalog No. 50-005-15703 Supplier MSC Supplier No. 99592404
This clamshell hard case provides robust protection for equipment. Constructed from durable polypropylene with layered foam, it measures 20-15/32 inches wide and is black. It's crush-proof, watertight, dust-proof, and IP67 rated. Features include an automatic pressure equalization valve and polyurethane wheels for easy transport.
  • Layered foam for customizable protection.
  • Padlockable design for secure storage.
  • Folding handles enable easy carrying.
  • Durable polypropylene construction.
  • IP67 rated for water and dust resistance.
  • Withstands -40°F to 210°F.
  • Crush-proof, solid wall design.
  • O-ring seal ensures watertight integrity.
  • Automatic pressure equalization valve.
  • Lightweight with polyurethane wheels.