Learn More
MSC Pelican Cube Case, Polypropylene, Watertight, Crushproof
Catalog No. 5000512653
Encompass
The Pelican Products, Inc. Cube Case is a robust storage solution crafted from durable polypropylene. Designed to protect sensitive equipment, it offers watertight, crushproof, and dustproof capabilities, making it suitable for various demanding environments.
  • Watertight design with O-ring seal
  • Crushproof solid wall construction
  • Dustproof protection for contents
  • Automatic pressure equalization valve
  • Lightweight yet strong open cell core
  • Meets STANAG 4280, IP67, and DEF STAN 81-41 standards
  • Made from polypropylene material
  • Padlockable for added security
  • Features a folding handle
Catalog No. 50-005-12653 Supplier MSC Supplier No. 77859320
The Pelican Products, Inc. Cube Case is a robust storage solution crafted from durable polypropylene. Designed to protect sensitive equipment, it offers watertight, crushproof, and dustproof capabilities, making it suitable for various demanding environments.
  • Watertight design with O-ring seal
  • Crushproof solid wall construction
  • Dustproof protection for contents
  • Automatic pressure equalization valve
  • Lightweight yet strong open cell core
  • Meets STANAG 4280, IP67, and DEF STAN 81-41 standards
  • Made from polypropylene material
  • Padlockable for added security
  • Features a folding handle