Learn More
MSC Pelican Micro Case, Polycarbonate, Watertight, IP67
Catalog No. 5000530410
Encompass
This micro case protects small sensitive components and smartphones from the elements. It features a crushproof, watertight, and dustproof design, capable of being submerged under 1 meter of water for up to 30 minutes, meeting IP67 testing parameters.
  • Crushproof, watertight, and dustproof construction
  • Rubber liner for extra protection also acts as an O-ring seal
  • Easy open latch for quick access
  • Integrated carabiner for portability
  • Automatic pressure equalization valve balances interior pressure
  • Submersible under 1 meter of water for up to 30 minutes
  • Available in clear with color liner or solid options
  • Lifetime guarantee (where applicable by law)
Catalog No. 50-005-30410 Supplier MSC Supplier No. 84765940
This micro case protects small sensitive components and smartphones from the elements. It features a crushproof, watertight, and dustproof design, capable of being submerged under 1 meter of water for up to 30 minutes, meeting IP67 testing parameters.
  • Crushproof, watertight, and dustproof construction
  • Rubber liner for extra protection also acts as an O-ring seal
  • Easy open latch for quick access
  • Integrated carabiner for portability
  • Automatic pressure equalization valve balances interior pressure
  • Submersible under 1 meter of water for up to 30 minutes
  • Available in clear with color liner or solid options
  • Lifetime guarantee (where applicable by law)