Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ MityLite™ Flashlights

Catalog No. 17985405E Shop All Pelican Products Inc. Products
Encompass_Preferred
Change view
Click to view available options
Color:
Yellow
Length (English):
4.09 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Length (English)
17-985-405E Yellow 4.09 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-985-405E Supplier Pelican™ Supplier No. 1900C YLW

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Beam cuts through thick smoke, fog, and dust

  • 1.5 hours burn time
  • High-intensity Xenon™ Laser Spot™ lamp produces a 11 Lumens beam
  • Xenoy polymer resin body with twist-on shroud is submersible and IPX7 rating

Order Info

Length: 4.1 in. (10.4cm). Weight (with batteries): 1.6 oz. (45.4g). Two AAA alkaline batteries.

Certifications

CSA approved for certain Class I, Division 2 hazardous locations; CE, IEC, and ETL marked.

Specifications

Gas Fill Xenon
Voltage 3 V
Weight (English) 1.6 oz.
Weight (Metric) 0.05 kg
Battery Run Time 1.5 hr.
Battery Type Alkaline
Certifications/Compliance CSA, CE, IEC, and ETL
Color Yellow
For Use With (Application) Uses a hi-intensity xenon lamp that produces a powerful, bright beam, beam cuts through thick smoke, fog, and dust
Includes Two AAA alkaline batteries
Length (English) 4.09 in.
Length (Metric) 10.4 cm
Lumen 11 lm
Material Polymer Resin
Product Type Pocket Light
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.