Learn More
MSC Pelican Products Aircase, Layered Foam, 9-1/8 in Deep, Polypropylene Blend, Black
Catalog No. 5000528650
Encompass
This aircase with layered foam is designed for robust protection in demanding environments. Constructed from a polypropylene blend, it features a depth of 9-1/8 in and comes in a black finish. It is built to be crush-proof, watertight, and dust-proof, meeting IP67 standards, and can withstand temperatures from -60°F (-51°C) to 160°F (71°C).
  • Layered foam for customizable protection
  • Business card holder
  • Automatic purge valve
  • Crush-proof construction
  • Watertight and dust-proof (IP67 certified)
  • Super-light proprietary design
  • Includes foam for immediate use
Catalog No. 50-005-28650 Supplier MSC Supplier No. 36003507
This aircase with layered foam is designed for robust protection in demanding environments. Constructed from a polypropylene blend, it features a depth of 9-1/8 in and comes in a black finish. It is built to be crush-proof, watertight, and dust-proof, meeting IP67 standards, and can withstand temperatures from -60°F (-51°C) to 160°F (71°C).
  • Layered foam for customizable protection
  • Business card holder
  • Automatic purge valve
  • Crush-proof construction
  • Watertight and dust-proof (IP67 certified)
  • Super-light proprietary design
  • Includes foam for immediate use