Learn More
MSC Pelican Products Clamshell Hard Case, 7-51/64 in Wide, Watertight, Crush Proof
Catalog No. 5000513683
Encompass
This clamshell hard case, constructed from durable plastic, is designed to protect sensitive equipment. It features a robust solid wall design and measures approximately 7-51/64 inches wide. Ideal for various applications requiring secure storage and transport.
  • Solid wall design.
  • Crush proof.
  • O-ring seal.
  • Automatic pressure equalization valve.
  • Lightweight.
  • Watertight.
  • Open cell core.
  • Dust proof.
  • Strong.
  • Includes foam insert.
Catalog No. 50-005-13683 Supplier MSC Supplier No. 67122499
This clamshell hard case, constructed from durable plastic, is designed to protect sensitive equipment. It features a robust solid wall design and measures approximately 7-51/64 inches wide. Ideal for various applications requiring secure storage and transport.
  • Solid wall design.
  • Crush proof.
  • O-ring seal.
  • Automatic pressure equalization valve.
  • Lightweight.
  • Watertight.
  • Open cell core.
  • Dust proof.
  • Strong.
  • Includes foam insert.