Learn More
MSC Pelican Products Clamshell Hard Case, Layered Foam, Watertight, Polypropylene
Catalog No. 5000514946
Encompass
This clamshell hard case features a durable polypropylene construction and layered foam interior, providing robust protection for sensitive equipment. Designed to be watertight, dustproof, and crush proof, it ensures the safety of contents in challenging environments. The case also includes an automatic pressure equalization valve and polyurethane wheels for convenient handling and transport.
  • Crush proof design
  • Watertight and dust proof protection
  • O-ring seal
  • Automatic pressure equalization valve
  • Lightweight yet strong construction
  • Polyurethane wheels for easy transport
  • Padlockable for security
  • Folding handle
  • Includes layered foam
  • Stainless steel bearings
Catalog No. 50-005-14946 Supplier MSC Supplier No. 79597993
This clamshell hard case features a durable polypropylene construction and layered foam interior, providing robust protection for sensitive equipment. Designed to be watertight, dustproof, and crush proof, it ensures the safety of contents in challenging environments. The case also includes an automatic pressure equalization valve and polyurethane wheels for convenient handling and transport.
  • Crush proof design
  • Watertight and dust proof protection
  • O-ring seal
  • Automatic pressure equalization valve
  • Lightweight yet strong construction
  • Polyurethane wheels for easy transport
  • Padlockable for security
  • Folding handle
  • Includes layered foam
  • Stainless steel bearings