Learn More
MSC Pelican Products Shipping Case, Layered Foam, Watertight, 34 x 24 x 12.5 in
Catalog No. 5000536952
Encompass
This shipping case provides robust protection for sensitive equipment with its solid wall design, crush-proof construction, and O-ring seal. Featuring layered foam for customizable interior protection, it is also watertight and dustproof, making it ideal for demanding environments.
  • Solid wall design for enhanced durability.
  • Crush-proof to protect contents from impact.
  • O-ring seal ensures a tight, protective closure.
  • Automatic pressure equalization valve.
  • Lightweight for easier transport.
  • Watertight and dustproof to keep contents safe.
  • Open cell core provides strength without added weight.
  • Includes layered foam for customizable interior organization.
  • Padlockable for added security.
  • Constructed from durable polypropylene material.
  • Meets IP67 standards.
  • Withstands temperatures from -40°F to 210°F.
Catalog No. 50-005-36952 Supplier MSC Supplier No. 42005066
This shipping case provides robust protection for sensitive equipment with its solid wall design, crush-proof construction, and O-ring seal. Featuring layered foam for customizable interior protection, it is also watertight and dustproof, making it ideal for demanding environments.
  • Solid wall design for enhanced durability.
  • Crush-proof to protect contents from impact.
  • O-ring seal ensures a tight, protective closure.
  • Automatic pressure equalization valve.
  • Lightweight for easier transport.
  • Watertight and dustproof to keep contents safe.
  • Open cell core provides strength without added weight.
  • Includes layered foam for customizable interior organization.
  • Padlockable for added security.
  • Constructed from durable polypropylene material.
  • Meets IP67 standards.
  • Withstands temperatures from -40°F to 210°F.