Learn More
MSC Pelican Protective Case, Watertight, Crush Proof, HPX Resin, Layered Foam
Catalog No. 5000533656
Encompass
This protective case offers superior protection for your valuable equipment. Constructed from durable HPX High Performance Resin, it is designed to be watertight, crush proof, and dust proof, ensuring your items remain safe in various environments. The case comes with layered foam for customizable interior organization and features a secure press and pull latch system.
  • Watertight, crush proof, and dust proof design for ultimate protection.
  • Constructed from lightweight and durable HPX High Performance Resin.
  • Includes layered foam for custom fit and equipment security.
  • Equipped with a dependable press and pull latch system.
  • Features a comfortable soft grip, double-layered handle.
  • Integrated automatic pressure equalization valve.
  • Certified to MIL-STD-810F, FED-STD-101C, and ATA 300 standards.
  • Withstands temperatures from -20°F to 140°F.
Catalog No. 50-005-33656 Supplier MSC Supplier No. 30619738
This protective case offers superior protection for your valuable equipment. Constructed from durable HPX High Performance Resin, it is designed to be watertight, crush proof, and dust proof, ensuring your items remain safe in various environments. The case comes with layered foam for customizable interior organization and features a secure press and pull latch system.
  • Watertight, crush proof, and dust proof design for ultimate protection.
  • Constructed from lightweight and durable HPX High Performance Resin.
  • Includes layered foam for custom fit and equipment security.
  • Equipped with a dependable press and pull latch system.
  • Features a comfortable soft grip, double-layered handle.
  • Integrated automatic pressure equalization valve.
  • Certified to MIL-STD-810F, FED-STD-101C, and ATA 300 standards.
  • Withstands temperatures from -20°F to 140°F.