Learn More
MSC Pelican Protector Case, Polypropylene, Watertight, IP67
Catalog No. 5000514862
Encompass
Designed for sensitive equipment, these rugged cases endure the harshest environments. Manufactured in the USA, they feature an automatic purge valve to balance air pressure, a watertight silicone O-ring lid, over-molded rubber handles, and stainless steel hardware.
  • Pick N Pluck™ with convoluted lid foam
  • Watertight, crushproof, and dustproof
  • Open cell core with solid wall design for lightweight strength
  • Automatic pressure equalization valve
  • Legendary lifetime guarantee
  • O-ring seal and stainless steel hardware
  • Meets rigorous US/NATO mil-spec certifications
  • Ultra-reliable Double Throw latches
Catalog No. 50-005-14862 Supplier MSC Supplier No. 00544585
Designed for sensitive equipment, these rugged cases endure the harshest environments. Manufactured in the USA, they feature an automatic purge valve to balance air pressure, a watertight silicone O-ring lid, over-molded rubber handles, and stainless steel hardware.
  • Pick N Pluck™ with convoluted lid foam
  • Watertight, crushproof, and dustproof
  • Open cell core with solid wall design for lightweight strength
  • Automatic pressure equalization valve
  • Legendary lifetime guarantee
  • O-ring seal and stainless steel hardware
  • Meets rigorous US/NATO mil-spec certifications
  • Ultra-reliable Double Throw latches