Learn More
MSC Pelican Storm Case, HPX Resin, Watertight, 15 x 10.5 x 6 in
Catalog No. 5000531072
Encompass
The Storm Case is a durable protective case designed to safeguard your gear. Constructed from injection molded HPX high performance resin, it combines lightweight design with rugged durability. It is built to protect against the harshest conditions, offering watertight, crushproof, and dustproof protection with interior dimensions of 15.00 x 10.50 x 6.00 inches.
  • Two Press & Pull latches
  • Lightweight strong HPX resin
  • Watertight, crushproof, and dustproof protection
  • Vortex valve
  • Double-layered, soft-grip handle
  • Two padlockable hasps
  • Powerful hinges
  • Guaranteed for life
  • Military-grade protection
  • Includes foam
Catalog No. 50-005-31072 Supplier MSC Supplier No. 87609103
Add to Cart
Add to Cart
The Storm Case is a durable protective case designed to safeguard your gear. Constructed from injection molded HPX high performance resin, it combines lightweight design with rugged durability. It is built to protect against the harshest conditions, offering watertight, crushproof, and dustproof protection with interior dimensions of 15.00 x 10.50 x 6.00 inches.
  • Two Press & Pull latches
  • Lightweight strong HPX resin
  • Watertight, crushproof, and dustproof protection
  • Vortex valve
  • Double-layered, soft-grip handle
  • Two padlockable hasps
  • Powerful hinges
  • Guaranteed for life
  • Military-grade protection
  • Includes foam