Learn More
MSC Pelican Storm Travel Case, Waterproof, Dustproof, Crushproof, Extendable Handle
Catalog No. 5000510192
Encompass
This travel case features unique Press and Pull latches that lock automatically and open with a light touch. It is guaranteed for life and engineered with the finest components. Designed for military-grade protection, it combines a lightweight design with rugged durability to meet mil-spec standards. It is waterproof, dustproof, and crushproof, built to protect gear against the harshest conditions.
  • Unique press and pull latches that lock automatically
  • Guaranteed for life
  • Two double-layered, soft-grip handles
  • Two padlockable hasps
  • Powerful hinges
  • O-ring seal
  • Military-grade protection
  • Waterproof, dustproof, crushproof construction
  • Extendable handle
Catalog No. 50-005-10192 Supplier MSC Supplier No. 92069806
This travel case features unique Press and Pull latches that lock automatically and open with a light touch. It is guaranteed for life and engineered with the finest components. Designed for military-grade protection, it combines a lightweight design with rugged durability to meet mil-spec standards. It is waterproof, dustproof, and crushproof, built to protect gear against the harshest conditions.
  • Unique press and pull latches that lock automatically
  • Guaranteed for life
  • Two double-layered, soft-grip handles
  • Two padlockable hasps
  • Powerful hinges
  • O-ring seal
  • Military-grade protection
  • Waterproof, dustproof, crushproof construction
  • Extendable handle