Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pelican™ Super SabreLite™ Flashlights

Catalog No. 19816476 Shop All Pelican Products Inc. Products
Change view
Click to view available options
Color:
Clear
Product Type:
Replacement Lens
Material:
Polycarbonate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Product Type Material
19-816-476 Clear Replacement Lens Polycarbonate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-816-476 Supplier Pelican™ Supplier No. 2003921110

missing translation for 'validMainframeMsg'

Includes

Spring clip, ring, and lanyard.

Requires

Three C alkaline batteries (not included).

Suitable for potentially explosive and other hazardous environments

Model available with photoluminescent shroud.

  • High-intensity Xenon Laser Spot™ lamp module 109 Lumens
  • Unbreakable ABS plastic-resin body and Lexan™ lens
  • IPX7 Rating: waterproof (submerged for 1m for 30 minutes)

Photoluminescent shroud

  • Lens ring emits a bright glow in the dark
  • Material in plastic lens ring absorbs energy from any light source: fluorescent lights, the sun, even its own beam

Order Info

Length: 7.79 in. (19.8cm). Weight: 12.8 oz. (362g).

Certifications

UL listed for Class 1. Division 1; Group C and D

Specifications

Battery Type 3 C Alkaline
Certifications/Compliance UL, Class 1. Division 1, Group C and D, ANSI/NEMA FL 1
Color Clear
For Use With (Application) Suitable for potentially explosive and other hazardous environments
For Use With (Equipment) SabreLite™ 2000 flashlight
Material Polycarbonate
Product Type Replacement Lens
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.