Learn More
Selleck Chemical LLC Pelitinib (EKB-569) 5mg 257933-82-7
SDP
Supplier:  Selleck Chemical LLC S13925MG
Encompass_Preferred
Pelitinib (EKB-569) is a potent irreversible EGFR inhibitor with IC50 of 38.5 nM. Pelitinib (EKB-569) also slightly inhibits Src, MEK/ERK and ErbB2 with IC50s of 282 nM, 800 nM and 1255 nM, respectively. Phase2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-801-8