Learn More
Abcam Pelitinib (EKB-569), Irreversible EGFR inhibitor, 5MG
Catalog No. 502706835
Encompass_Preferred
MW 467.9 Da, Purity >97%. Irreversible EGFR inhibitor (IC₅₀ = 38.5 nM). Greater inhibitory activity against EGFR then erbB-2, Src, Cdk4, c-Met, Raf, MEK/ERK (IC₅₀ values are 1255, 282, >20,000, 4100, 3353 and 800 nM respectively). Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6835 Supplier Abcam Supplier No. AB1411715MG
MW 467.9 Da, Purity >97%. Irreversible EGFR inhibitor (IC₅₀ = 38.5 nM). Greater inhibitory activity against EGFR then erbB-2, Src, Cdk4, c-Met, Raf, MEK/ERK (IC₅₀ values are 1255, 282, >20,000, 4100, 3353 and 800 nM respectively). Active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement