Learn More
Warner Instruments™ Pellet Electrode
Catalog No. BTX641309 Shop All BTX Harvard Products
Encompass_Preferred
Change view
Click to view available options
Type:
Pellet Electrode
Catalog No. Type
BTX641309 Pellet Electrode
1 options
Catalog No. BTX641309 Supplier Warner Instruments™ Supplier No. 641309
A 1.0 mm pellet and wire electrode made from high-purity Ag or Ag/AgCl supports tissue probing and bioelectrical recordings.
  • Pellet electrode (E205)
  • The E205 pellet electrode has a cylindrical AgCl pellet embedded on a 0.25 x 70 mm wire.
  • Suitable for probing tissue
  • Has a 1.0 mm diameter Ag/AgCl matrix at the tip
Specifications
Type Pellet Electrode