Learn More
MSC Bare Ground Solutions Ice Melter, Pellets, 2,000 lb, Skidded Supersack
Catalog No. 502836811
Encompass
This ice and snow melter and de-icer comes in a pellet form, packaged in a 2,000 lb skidded supersack. Formulated with calcium chloride, it is effective down to -20 °F and is environmentally safe. The product also includes traction granules to enhance safety on icy surfaces and is biodegradable.
  • Pellet form for efficient spreading
  • Environmentally safe and biodegradable formula
  • Includes traction granules for enhanced safety
  • Effective at temperatures as low as -20 °F
  • Composed of calcium chloride
  • Packaged in a convenient 2,000 lb skidded supersack
Catalog No. 50-283-6811 Supplier MSC Supplier No. 55598783
This ice and snow melter and de-icer comes in a pellet form, packaged in a 2,000 lb skidded supersack. Formulated with calcium chloride, it is effective down to -20 °F and is environmentally safe. The product also includes traction granules to enhance safety on icy surfaces and is biodegradable.
  • Pellet form for efficient spreading
  • Environmentally safe and biodegradable formula
  • Includes traction granules for enhanced safety
  • Effective at temperatures as low as -20 °F
  • Composed of calcium chloride
  • Packaged in a convenient 2,000 lb skidded supersack