Learn More
Sigma Aldrich Fine Chemicals Biosciences Pellet pestles Cordless mo1EA
Catalog No. 502900351
Encompass
Z359955 will fit the cordless motor product Z359971 only by using the replacement adapter for the motor Z359998 which is purchased separately.
Catalog No. 50-290-0351 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z3599711EA
Z359955 will fit the cordless motor product Z359971 only by using the replacement adapter for the motor Z359998 which is purchased separately.