Learn More
MSC J.T. Eaton Rodent Pellet Bait, Bromethalin, 128 Pk.
Catalog No. 502866684
Encompass
This pellet bait holder is designed for rodent control, specifically targeting mice and rats. It comes in 128 individual packs, each containing 0.5 oz of bait.
  • Form: pellet
  • Targets rats and mice
  • For rodent application
  • Active ingredients: sucrose and bromethalin
Catalog No. 50-286-6684 Supplier MSC Supplier No. 70698261
This pellet bait holder is designed for rodent control, specifically targeting mice and rats. It comes in 128 individual packs, each containing 0.5 oz of bait.
  • Form: pellet
  • Targets rats and mice
  • For rodent application
  • Active ingredients: sucrose and bromethalin