Learn More
MSC Peladow 28-6250 50 Lb Bag Environmentally Safe Pellet-Form Ice & Snow Melter & De-Icer Effective to -25°F, Calcium Chloride
Catalog No. 501305400
Encompass
Peladow 28-6250 50 Lb Bag Environmentally Safe Pellet-Form Ice & Snow Melter & De-Icer Effective to -25°F, Calcium Chloride
Catalog No. 50-130-5400 Supplier MSC Supplier No. 09246182
Peladow 28-6250 50 Lb Bag Environmentally Safe Pellet-Form Ice & Snow Melter & De-Icer Effective to -25°F, Calcium Chloride