Learn More
MilliporeSigma Pellicon 3 Cassette with Ultracel 30 kDa Membrane, C screen, 88 cm
Supplier:  MilliporeSigma P3C030C00
Encompass
Pellicon 3 Cassette w/Ultracel PLCTK Membrane, 88 cm2. Device Configuration: Cassette. Filter Code: PLCTK. Maximum Inlet Pressure, bar (psi): 2.75 bar (40 psi) @ 50 C; 5.5 bar (80 psi) @ 40 C. Maximum Transmembrane Pressure: 5.5 bar (80 psig) @ 30 C. Filter Type: Ultrafiltration. Good Manufacturing Practices: These products are manufactured in a facility which adheres to FDA Good Manufacturing Practices. NMWL: 30 kDa. Key Applications are Protein Purification and Analytical Sample Prep.
Catalog No. NC0865632