Learn More
MilliporeSigma Pellicon 3 Cassette with Ultracel Membrane
Supplier:  MilliporeSigma P3C005C01
Encompass
Filtration area 0.11 m2, NMWCO 10 kDa. Type C screen (coarse screen).
Catalog No. NC0794386