Learn More
MilliporeSigma Pellicon 3 Cassette with Ultracel Membrane
Supplier:  MilliporeSigma P3C003C00
Encompass
Pellicon 3 Cassette with Ultracel 3kDa Membrane, C Screen, 88 cm2.
Catalog No. NC0784306