Learn More
Cayman Chemical Pellitorine 10mg
SDP
Catalog No. 502952526
Encompass_Preferred
A partal agonst of PPARY actvates PPARY wth a lower potency (K 15 versus 0 035 UM) but a smlar maxmal effcacy compared to rosgltazone mproves nsuln resstance n normal detnduced glucosentolerant and n dabetc db/db mce yet reduced adpogenc actvty
Catalog No. 50-295-2526 Supplier Cayman Chemical Supplier No. 11662 10MG
A partal agonst of PPARY actvates PPARY wth a lower potency (K 15 versus 0 035 UM) but a smlar maxmal effcacy compared to rosgltazone mproves nsuln resstance n normal detnduced glucosentolerant and n dabetc db/db mce yet reduced adpogenc actvty