Learn More
Leica Biosystems Peloris Parablocks X-tra (5 Bl
Catalog No. 502552381
Encompass_Preferred
The preformed blocks fit easily into the paraffin chamber of the Leica PELORIS Tissue Processor and will melt faster than pellets or flakes.Once the chamber is filled with the solid paraffin blocks, there is no need for the technician to return later to determine the paraffin volume and add additional paraffin.May reduce risks since solid blocks are designed to help prevent pellet spills and the need to transfer molten paraffin.Unique to PELORIS Tissue Processors – designed to suit the needs of PELORIS paraffin chambers.Promote Lab Safety – Solid blocks help prevent pellet spills and the need to transfer molten paraffin.Easy Loading – Parablocks ensures the correct amount of paraffin is loaded every time and is less temperature sensitive so there is no top off during melting and less waste.
This item is not returnable. View return policy
Catalog No. 50-255-2381 Supplier Leica Biosystems Supplier No. 39V3001
The preformed blocks fit easily into the paraffin chamber of the Leica PELORIS Tissue Processor and will melt faster than pellets or flakes.Once the chamber is filled with the solid paraffin blocks, there is no need for the technician to return later to determine the paraffin volume and add additional paraffin.May reduce risks since solid blocks are designed to help prevent pellet spills and the need to transfer molten paraffin.Unique to PELORIS Tissue Processors – designed to suit the needs of PELORIS paraffin chambers.Promote Lab Safety – Solid blocks help prevent pellet spills and the need to transfer molten paraffin.Easy Loading – Parablocks ensures the correct amount of paraffin is loaded every time and is less temperature sensitive so there is no top off during melting and less waste.