Learn More
TA Instruments Peltier Concentric Cylinder Solvent Trap, for Standard 30 mm Diameter Cup, Compatible with Conical DIN, Recessed End, Vaned Rotors, Not for Double Wall System or Large Diameter Peltier Cup
Catalog No. 5000580880 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Peltier Concentric Cylinder Solvent Trap is designed for use with standard 30 mm diameter cups and is compatible with conical DIN, recessed end, and vaned rotors. It is not for use with double wall systems or large diameter Peltier cups.
Catalog No. 50-005-80880 Supplier TA Instruments Supplier No. 545609.901
Waters TA Instruments Peltier Concentric Cylinder Solvent Trap is designed for use with standard 30 mm diameter cups and is compatible with conical DIN, recessed end, and vaned rotors. It is not for use with double wall systems or large diameter Peltier cups.